siRNA Details
Gene Symbol: ZNF169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301275.1 | NP_001288204.1 | 5 | 886 | ||
NM_003448.2 | NP_003439.2 | 5 | 522 | |||
NM_194320.3 | NP_919301.2 | 5 | 519 | |||
XM_017014362.1 | XP_016869851.1 | 5 | 651 | |||
XM_017014363.1 | XP_016869852.1 | 6 | 545 | |||
XM_017014364.1 | XP_016869853.1 | 6 | 542 | |||
XM_017014365.1 | XP_016869854.1 | 5 | 6875 | |||
XM_017014366.1 | XP_016869855.1 | 5 | 6813 | |||
GenBank mRNA | AK093956.1 | - | 5 | 824 | ||
AK122832.1 | - | 1 | 1142 | |||
AK289742.1 | - | 5 | 463 | |||
BC047702.2 | - | 4 | 443 | 5585128 | ||
BP873364.1 | - | 5 | 474 | |||
DA490480.1 | - | 5 | 490 |
