siRNA Details
Gene Symbol: FAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000043.5 | NP_000034.1 | 9 | 1065 | ||
NM_001320619.1 | NP_001307548.1 | 8 | 982 | |||
NM_152871.3 | NP_690610.1 | 8 | 1002 | |||
NM_152872.3 | NP_690611.1 | 8 | 1040 | |||
NR_028033.3 | 7 | 893 | ||||
NR_028034.3 | 6 | 755 | ||||
NR_028035.3 | 7 | 818 | ||||
NR_028036.3 | 8 | 956 | ||||
NR_135313.1 | 7 | 873 | ||||
NR_135314.1 | 9 | 1056 | ||||
NR_135315.1 | 7 | 809 | ||||
XM_006717819.3 | XP_006717882.1 | 10 | 5747 | |||
XM_011539764.2 | XP_011538066.1 | 10 | 1136 | |||
XM_011539765.2 | XP_011538067.1 | 9 | 861 | |||
XM_011539766.2 | XP_011538068.1 | 10 | 876 | |||
XM_011539767.2 | XP_011538069.1 | 9 | 799 | |||
GenBank mRNA | AB209361.1 | - | 10 | 4382 | ||
AK290978.1 | - | 9 | 993 | |||
AV715411.1 | - | 7 | 654 | |||
AY495076.2 | - | 9 | 941 | |||
BC012479.1 | - | 9 | 1057 | 4514272 | ||
CR450307.1 | - | 9 | 747 | |||
EU481974.2 | - | 9 | 942 | |||
FJ200481.1 | - | 8 | 664 | |||
FJ373046.1 | - | 9 | 746 | |||
FM246458.1 | - | 9 | 747 | |||
FM246459.1 | - | 9 | 748 | |||
M67454.1 | - | 9 | 941 | |||
X63717.1 | - | 9 | 967 | |||
X83490.1 | - | 7 | 525 | |||
X83491.1 | - | 6 | 462 | |||
X83492.1 | - | 7 | 580 | |||
X83493.1 | - | 8 | 709 | |||
Z47993.1 | - | 8 | 684 | |||
Z47994.1 | - | 7 | 500 | |||
Z47995.1 | - | 6 | 437 | |||
Z66556.1 | - | 8 | 722 | |||
Z70519.1 | - | 8 | 638 | |||
Z70520.1 | - | 7 | 575 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
