siRNA Details
Gene Symbol: CD36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000072.3 | NP_000063.2 | 3 | 492 | ||
NM_001001547.2 | NP_001001547.1 | 3 | 453 | |||
NM_001001548.2 | NP_001001548.1 | 3 | 492 | |||
NM_001127443.1 | NP_001120915.1 | 2 | 198 | |||
NM_001127444.1 | NP_001120916.1 | 1 | 373 | |||
NM_001289908.1 | NP_001276837.1 | 1 | 166 | |||
NM_001289909.1 | NP_001276838.1 | 2 | 257 | |||
NR_110501.1 | 2 | 257 | ||||
XM_005250713.1 | XP_005250770.1 | 3 | 434 | |||
XM_005250714.1 | XP_005250771.1 | 3 | 401 | |||
XM_005250715.4 | XP_005250772.1 | 3 | 559 | |||
GenBank mRNA | AK096858.1 | - | 6 | 762 | ||
AK129899.1 | - | 3 | 434 | |||
AK312920.1 | - | 3 | 416 | |||
AL596924.1 | - | 1 | 377 | |||
BC008406.1 | - | 3 | 367 | 4244251 | ||
BP258762.1 | - | 3 | 453 | |||
BP259746.1 | - | 3 | 452 | |||
DA566475.1 | - | 2 | 198 | |||
DC338512.1 | - | 3 | 393 | |||
HM217020.1 | - | 1 | 175 | |||
HM217021.1 | - | 1 | 175 | |||
HM217022.1 | - | 1 | 175 | |||
HM217023.1 | - | 1 | 175 | |||
HM217024.1 | - | 1 | 175 | |||
HM217025.1 | - | 1 | 175 | |||
HM217026.1 | - | 1 | 166 | |||
KU177961.1 | - | 1 | 78 | |||
KU177962.1 | - | 1 | 78 | |||
KU177963.1 | - | 1 | 78 | |||
L06850.1 | - | 2 | 210 | |||
M24795.1 | - | 3 | 288 | |||
M98398.1 | - | 3 | 332 | |||
M98399.1 | - | 3 | 332 | |||
S67532.1 | - | 2 | 214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
