siRNA Details
Gene Symbol: CD36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000072.3 | NP_000063.2 | 10 | 1281 | ||
NM_001001547.2 | NP_001001547.1 | 10 | 1242 | |||
NM_001001548.2 | NP_001001548.1 | 10 | 1281 | |||
NM_001127443.1 | NP_001120915.1 | 9 | 987 | |||
NM_001127444.1 | NP_001120916.1 | 8 | 1162 | |||
NM_001289908.1 | NP_001276837.1 | 6 | 838 | |||
NM_001289909.1 | NP_001276838.1 | 8 | 866 | |||
NM_001289911.1 | NP_001276840.1 | 8 | 920 | |||
XM_005250713.1 | XP_005250770.1 | 10 | 1223 | |||
XM_005250714.1 | XP_005250771.1 | 10 | 1190 | |||
XM_005250715.4 | XP_005250772.1 | 10 | 1348 | |||
GenBank mRNA | AK096858.1 | - | 13 | 1551 | ||
AK129899.1 | - | 10 | 1223 | |||
AK300101.1 | - | 8 | 920 | |||
AK312920.1 | - | 10 | 1205 | |||
BC008406.1 | - | 10 | 1156 | 4244251 | ||
CR749465.1 | - | 2 | 1649 | |||
HM217020.1 | - | 8 | 964 | |||
HM217021.1 | - | 8 | 964 | |||
HM217022.1 | - | 8 | 964 | |||
HM217023.1 | - | 7 | 784 | |||
HM217024.1 | - | 8 | 964 | |||
HM217026.1 | - | 6 | 838 | |||
KU177961.1 | - | 8 | 867 | |||
L06850.1 | - | 9 | 999 | |||
M24795.1 | - | 10 | 1077 | |||
M98398.1 | - | 10 | 1121 | |||
M98399.1 | - | 10 | 1121 | |||
S67532.1 | - | 9 | 1003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
