siRNA Details
Gene Symbol: MS4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000139.4 | NP_000130.1 | 3 | 377 | ||
XM_005273846.4 | XP_005273903.1 | 4 | 692 | |||
XM_011544850.2 | XP_011543152.1 | 4 | 310 | |||
XM_017017362.1 | XP_016872851.1 | 4 | 692 | |||
GenBank mRNA | AK304487.1 | - | 3 | 377 | ||
AK310142.1 | - | 3 | 377 | |||
BC074800.2 | - | 3 | 306 | 30915525 | ||
BC074843.2 | - | 3 | 306 | 30915222 | ||
D10583.1 | - | 3 | 325 | |||
DA972757.1 | - | 3 | 377 | |||
X66187.1 | - | 1 | 65 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
