siRNA Details
Gene Symbol: FUT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000149.3 | NP_000140.1 | 3 | 2345 | ||
NM_001097639.1 | NP_001091108.1 | 3 | 1997 | |||
NM_001097640.1 | NP_001091109.1 | 3 | 1943 | |||
NM_001097641.1 | NP_001091110.1 | 2 | 1804 | |||
XM_011527865.1 | XP_011526167.1 | 3 | 2323 | |||
XM_011527866.1 | XP_011526168.1 | 3 | 1984 | |||
XM_011527867.1 | XP_011526169.1 | 3 | 1953 | |||
GenBank mRNA | AF131913.1 | - | 2 | 1876 | ||
U27326.1 | - | 3 | 1936 | |||
U27327.1 | - | 3 | 1990 | |||
U27328.1 | - | 4 | 2339 | |||
X53578.1 | - | 2 | 1779 |
