siRNA Details
Gene Symbol: CSF2RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000395.2 | NP_000386.1 | 14 | 2292 | ||
XM_005261340.3 | XP_005261397.1 | 14 | 2369 | |||
XM_011529903.2 | XP_011528205.1 | 14 | 2405 | |||
XM_011529904.2 | XP_011528206.1 | 14 | 2387 | |||
GenBank mRNA | AK302993.1 | - | 12 | 1954 | ||
BC070085.1 | - | 14 | 2292 | 30344148 | ||
CR456428.1 | - | 13 | 2166 | |||
M59941.1 | - | 13 | 2103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
