siRNA Details
Gene Symbol: IL4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000418.3 | NP_000409.1 | 6 | 650 | ||
NM_001257406.1 | NP_001244335.1 | 5 | 517 | |||
NM_001257407.1 | NP_001244336.1 | 6 | 761 | |||
NM_001257997.1 | NP_001244926.1 | 5 | 517 | |||
XM_011545825.1 | XP_011544127.1 | 5 | 555 | |||
XM_011545826.2 | XP_011544128.1 | 6 | 606 | |||
XM_011545827.2 | XP_011544129.1 | 6 | 1845 | |||
XM_011545828.2 | XP_011544130.1 | 4 | 323 | |||
XM_017023211.1 | XP_016878700.1 | 5 | 555 | |||
GenBank mRNA | AK223591.1 | - | 6 | 568 | ||
AK303255.1 | - | 6 | 736 | |||
AK314863.1 | - | 5 | 502 | |||
BC136365.1 | - | 5 | 549 | 9020352 | ||
BC151131.1 | - | 5 | 549 | 9057031 | ||
DA492909.1 | - | 5 | 518 | |||
X52425.1 | - | 6 | 562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
