siRNA Details
Gene Symbol: LDLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000527.4 | NP_000518.1 | 13, 14 | 2182 | ||
NM_001195798.1 | NP_001182727.1 | 13, 14 | 2182 | |||
NM_001195799.1 | NP_001182728.1 | 12, 13 | 2059 | |||
NM_001195800.1 | NP_001182729.1 | 11, 12 | 1678 | |||
XM_011528010.1 | XP_011526312.1 | 13, 14 | 2145 | |||
GenBank mRNA | AB209409.1 | - | 13, 14 | 2066 | ||
AK295612.1 | - | 12, 13 | 1718 | |||
AK299038.1 | - | 11, 12 | 1577 | |||
AK300313.1 | - | 12, 13 | 1958 | |||
AY114155.1 | - | 13, 14 | 2009 | |||
BC014514.1 | - | 13, 14 | 2065 | 3542845 | ||
BT007361.1 | - | 13, 14 | 1995 | |||
BX648281.1 | - | 13, 14 | 2250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
