siRNA Details
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000566.3 | NP_000557.1 | 3 | 153 | ||
XM_005244957.3 | XP_005245014.1 | 3 | 156 | |||
GenBank mRNA | AK291451.1 | - | 3 | 136 | ||
AK291502.1 | - | 3 | 153 | |||
BC032634.1 | - | 3 | 115 | 5248549 | ||
BC152383.1 | - | 3 | 106 | 100013442 | ||
DC320241.1 | - | 3 | 153 | |||
L03418.1 | - | 3 | 103 | |||
X14355.1 | - | 3 | 158 | |||
X14356.1 | - | 3 | 139 |
Gene Symbol: FCGR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017986.3 | NP_001017986.1 | 3 | 153 | ||
NM_001244910.1 | NP_001231839.1 | 3 | 153 | |||
NR_045213.1 | 3 | 153 | ||||
XM_017000661.1 | XP_016856150.1 | 3 | 156 | |||
GenBank mRNA | AK308886.1 | - | 3 | 139 | ||
AK311502.1 | - | 2 | 965 | |||
L03419.1 | - | 3 | 103 |
Gene Symbol: FCGR1CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027484.2 | 3 | 153 | |||
GenBank mRNA | T29148.1 | - | 3 | 92 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
