siRNA Details
Gene Symbol: FCGR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004340.3 | NP_001004340.2 | 3 | 1027 | ||
NM_001017986.3 | NP_001017986.1 | 5 | 1303 | |||
NR_045213.1 | 6 | 1588 | ||||
XM_017000661.1 | XP_016856150.1 | 5 | 1306 | |||
XM_017000662.1 | XP_016856151.1 | 4 | 1048 |
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000566.3 | NP_000557.1 | 6 | 1585 | ||
GenBank mRNA | AK291502.1 | - | 1641 | |||
BC032634.1 | - | 6 | 1547 | 5248549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
