siRNA Details
Gene Symbol: IFNAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000629.2 | NP_000620.2 | 3 | 503 | ||
XM_005260964.2 | XP_005261021.1 | 3 | 476 | |||
XM_011529552.1 | XP_011527854.1 | 3 | 503 | |||
GenBank mRNA | AK222770.1 | - | 3 | 439 | ||
AK222812.1 | - | 3 | 439 | |||
AK298051.1 | - | 3 | 490 | |||
AK312631.1 | - | 3 | 439 | |||
BC002590.2 | - | 3 | 418 | |||
BC021825.1 | - | 3 | 435 | 4932084 | ||
BG548299.1 | - | 3 | 508 | |||
J03171.1 | - | 3 | 434 | |||
JN543953.1 | - | 3 | 437 | |||
JN543954.1 | - | 3 | 437 | |||
JN543955.1 | - | 3 | 437 | |||
JN543956.1 | - | 3 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
