siRNA Details
Gene Symbol: IFNAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000629.2 | NP_000620.2 | 8 | 1244 | ||
XM_005260964.2 | XP_005261021.1 | 8 | 1217 | |||
GenBank mRNA | AK222770.1 | - | 8 | 1180 | ||
AK222812.1 | - | 8 | 1180 | |||
AK298051.1 | - | 8 | 1231 | |||
AK312631.1 | - | 8 | 1180 | |||
BC002590.2 | - | 8 | 1159 | |||
BC021825.1 | - | 8 | 1176 | 4932084 | ||
J03171.1 | - | 8 | 1175 | |||
JN543956.1 | - | 8 | 1178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
