siRNA Details
Gene Symbol: ITGAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000632.3 | NP_000623.2 | 5, 6 | 533 | ||
NM_001145808.1 | NP_001139280.1 | 5, 6 | 533 | |||
XM_006721045.1 | XP_006721108.1 | 5, 6 | 525 | |||
XM_011545850.2 | XP_011544152.1 | 3, 4 | 329 | |||
XM_011545851.2 | XP_011544153.1 | 5, 6 | 525 | |||
XM_017023216.1 | XP_016878705.1 | 5, 6 | 525 | |||
GenBank mRNA | BC096346.3 | - | 5, 6 | 499 | 40008964 | |
BC096347.3 | - | 5, 6 | 499 | 40008967 | ||
BC096348.3 | - | 5, 6 | 499 | 40008972 | ||
BC099660.3 | - | 5, 6 | 499 | 40008969 | ||
J03925.1 | - | 5, 6 | 507 | |||
J04145.1 | - | 4, 5 | 411 | |||
M18044.1 | - | 5, 6 | 510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
