siRNA Details
Gene Symbol: MASTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172303.2 | NP_001165774.1 | 3, 4 | 1075 | ||
NM_001172304.2 | NP_001165775.1 | 3, 4 | 1075 | |||
NM_001320756.1 | NP_001307685.1 | 3, 4 | 1075 | |||
NM_001320757.1 | NP_001307686.1 | 3, 4 | 1075 | |||
NM_032844.4 | NP_116233.2 | 3, 4 | 1075 | |||
NR_135469.1 | 3, 4 | 1075 | ||||
XM_005252631.3 | XP_005252688.1 | 3, 4 | 1428 | |||
XM_005252632.3 | XP_005252689.1 | 3, 4 | 1428 | |||
XM_006717519.3 | XP_006717582.1 | 3, 4 | 1428 | |||
XM_006717520.3 | XP_006717583.1 | 3, 4 | 1428 | |||
XM_017016852.1 | XP_016872341.1 | 3, 4 | 1428 | |||
XM_017016853.1 | XP_016872342.1 | 3, 4 | 1046 | |||
XM_017016854.1 | XP_016872343.1 | 3, 4 | 1428 | |||
XM_017016855.1 | XP_016872344.1 | 3, 4 | 1428 | |||
XM_017016856.1 | XP_016872345.1 | 3, 4 | 1428 | |||
GenBank mRNA | AK027719.1 | - | 3, 4 | 536 | ||
AK074804.1 | - | 3, 4 | 564 | |||
AK123004.1 | - | 3, 4 | 1977 | |||
AK225319.1 | - | 3, 4 | 505 | |||
BC009107.1 | - | 3, 4 | 529 | 3449913 | ||
DB150802.1 | - | 3, 4 | 547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
