siRNA Details
Gene Symbol: IL13RA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000640.2 | NP_000631.1 | 5 | 556 | ||
GenBank mRNA | AK291957.1 | - | 5 | 525 | ||
AK308523.1 | - | 5 | 487 | |||
AK309444.1 | - | 5 | 566 | |||
BC020739.1 | - | 4 | 457 | 4807603 | ||
BC033705.2 | - | 4 | 465 | 3876064 | ||
GQ494004.1 | - | 5 | 488 | |||
U70981.1 | - | 5 | 524 | |||
X95302.1 | - | 5 | 483 | |||
Y08768.1 | - | 5 | 556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
