siRNA Details
Gene Symbol: IGF2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000876.3 | NP_000867.2 | 18 | 2576 | ||
GenBank mRNA | AB209668.1 | - | 17 | 2199 | ||
J03528.1 | - | 18 | 2575 | |||
Y00285.1 | - | 18 | 2575 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
