siRNA Details
Gene Symbol: PAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002577.4 | NP_002568.2 | 2 | 409 | ||
XM_011512870.2 | XP_011511172.1 | 2 | 132 | |||
XM_017006501.1 | XP_016861990.1 | 3 | 596 | |||
GenBank mRNA | AK291339.1 | - | 2 | 293 | ||
BC069613.1 | - | 1 | 122 | 7262289 | ||
U24153.1 | - | 1 | 126 |
Gene Symbol: LOC646214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209428.1 | - | 1 | 2720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
