siRNA Details
Gene Symbol: P4HA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000917.3 | NP_000908.2 | 10, 11 | 1488 | ||
NM_001017962.2 | NP_001017962.1 | 10, 11 | 1488 | |||
NM_001142595.1 | NP_001136067.1 | 11, 12 | 1581 | |||
GenBank mRNA | AK222960.1 | - | 10, 11 | 1376 | ||
AK312593.1 | - | 10, 11 | 1374 | |||
BC034998.1 | - | 10, 11 | 1381 | 4797051 | ||
CD013930.1 | - | 11, 12 | 1460 | |||
M24486.1 | - | 10, 11 | 1365 | |||
M24487.1 | - | 10, 11 | 1365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
