siRNA Details
Gene Symbol: MAPK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002749.3 | NP_002740.2 | 6 | 2427 | ||
NM_139032.2 | NP_620601.1 | 4 | 2235 | |||
NM_139033.2 | NP_620602.2 | 6 | 2638 | |||
NM_139034.2 | NP_620603.2 | 6 | 2339 | |||
XM_006721557.3 | XP_006721620.1 | 7 | 2403 | |||
XM_006721558.3 | XP_006721621.1 | 7 | 2656 | |||
XM_006721559.3 | XP_006721622.1 | 7 | 2565 | |||
XM_011523957.2 | XP_011522259.1 | 4 | 2142 | |||
GenBank mRNA | AB209611.1 | - | 2 | 4027 | ||
AK295376.1 | - | 6 | 2332 | |||
AY534741.1 | - | 4 | 2359 | |||
BC007404.2 | - | 4 | 2203 | 3009873 | ||
BC007992.2 | - | 4 | 2203 | 3009873 | ||
BC009963.2 | - | 6 | 2359 | 4300124 | ||
BC030134.1 | - | 6 | 2274 | 4111084 | ||
U25278.1 | - | 6 | 2332 | |||
U29725.1 | - | 6 | 2473 | |||
U29726.1 | - | 6 | 2306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
