siRNA Details
Gene Symbol: PRLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000949.6 | NP_000940.1 | 4 | 558 | ||
NM_001204315.1 | NP_001191244.1 | 3 | 236 | |||
NM_001204316.1 | NP_001191245.1 | 3 | 236 | |||
NM_001204317.1 | NP_001191246.1 | 3 | 236 | |||
NM_001204318.1 | NP_001191247.1 | 3 | 236 | |||
XM_006714484.2 | XP_006714547.1 | 4 | 376 | |||
XM_011514068.2 | XP_011512370.1 | 4 | 391 | |||
XM_011514069.2 | XP_011512371.1 | 4 | 330 | |||
XM_017009646.1 | XP_016865135.1 | 2 | 174 | |||
GenBank mRNA | AF166329.1 | - | 2 | 131 | ||
AF416618.1 | - | 2 | 148 | |||
AF416619.1 | - | 2 | 148 | |||
AF492470.1 | - | 2 | 148 | |||
AK313270.1 | - | 4 | 446 | |||
BC059392.1 | - | 4 | 444 | 30343995 | ||
DA640476.1 | - | 4 | 391 | |||
GU133399.1 | - | 2 | 148 | |||
KJ651922.1 | - | 5 | 560 | |||
KJ651923.1 | - | 5 | 508 | |||
KJ651924.1 | - | 4 | 417 | |||
KJ651925.1 | - | 4 | 424 | |||
M31661.1 | - | 4 | 415 | |||
S78505.1 | - | 1 | 59 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
