siRNA Details
Gene Symbol: MAP3K14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003954.4 | NP_003945.2 | 2, 3 | 382 | ||
XM_011525441.2 | XP_011523743.1 | 3, 4 | 519 | |||
GenBank mRNA | AK290203.1 | - | 2, 3 | 358 | ||
AK313085.1 | - | 2, 3 | 358 | |||
BC035576.1 | - | 2, 3 | 327 | 5497185 | ||
DA856801.1 | - | 2, 3 | 366 | |||
HY096069.1 | - | 2, 3 | 385 | |||
Y10256.1 | - | 1, 2 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
