siRNA Details
Gene Symbol: MAP3K14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003954.4 | NP_003945.2 | 16 | 3630 | ||
XM_011525441.2 | XP_011523743.1 | 17 | 3767 | |||
GenBank mRNA | AK131438.1 | - | 8 | 2312 | ||
BC035576.1 | - | 16 | 3575 | 5497185 | ||
CR749592.1 | - | 13 | 3059 | |||
Y10256.1 | - | 15 | 3744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
