siRNA Details
Gene Symbol: STK17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004760.2 | NP_004751.2 | 2 | 451 | ||
XM_017012792.1 | XP_016868281.1 | 2 | 263 | |||
GenBank mRNA | AB011420.1 | - | 2 | 417 | ||
BC023508.2 | - | 2 | 407 | 3638004 | ||
BC047696.1 | - | 2 | 420 | 5761182 | ||
BI461892.1 | - | 2 | 484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
