siRNA Details
Gene Symbol: BRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113182.2 | NP_001106653.1 | 13 | 4359 | ||
NM_001199455.1 | NP_001186384.1 | 13 | 4154 | |||
NM_001199456.1 | NP_001186385.1 | 12 | 2762 | |||
NM_001291986.1 | NP_001278915.1 | 14 | 4451 | |||
NM_005104.3 | NP_005095.1 | 13 | 4446 | |||
GenBank mRNA | AL832722.1 | - | 14 | 4385 | ||
BC063840.1 | - | 13 | 3019 | 6181728 | ||
BX647233.1 | - | 12 | 2762 | |||
BX648109.1 | - | 13 | 4360 | |||
D42040.1 | - | 13 | 4446 | |||
M80613.1 | - | 12 | 3782 | |||
X62083.1 | - | 12 | 3782 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
