siRNA Details
Gene Symbol: AKT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014431.1 | NP_001014431.1 | 14 | 2398 | ||
NM_001014432.1 | NP_001014432.1 | 15 | 2482 | |||
NM_005163.2 | NP_005154.2 | 14 | 2612 | |||
XM_005267401.1 | XP_005267458.1 | 14 | 2547 | |||
GenBank mRNA | AK094287.1 | - | 2 | 1672 | ||
AK122894.1 | - | 14 | 3539 | |||
AK131465.1 | - | 2 | 2813 | |||
BC000479.2 | - | 14 | 2333 | 2964603 | ||
BC001737.1 | - | 5 | 1221 | |||
BC084538.1 | - | 15 | 2476 | 6645486 | ||
BX648205.1 | - | 13 | 3016 | |||
M63167.1 | - | 14 | 2256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
