siRNA Details
Gene Symbol: TNFRSF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192.2 | NP_001183.2 | 2 | 484 | ||
GenBank mRNA | EF152353.1 | - | 2 | 263 | ||
EF152354.1 | - | 2 | 173 | |||
EF152356.1 | - | 2 | 241 | |||
Z14954.1 | - | 2 | 332 | |||
Z29575.1 | - | 2 | 484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
