siRNA Details
Gene Symbol: EGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005228.3 | NP_005219.2 | 2 | 412 | ||
NM_201282.1 | NP_958439.1 | 2 | 412 | |||
NM_201283.1 | NP_958440.1 | 2 | 412 | |||
NM_201284.1 | NP_958441.1 | 2 | 412 | |||
GenBank mRNA | AB209442.1 | - | 2 | 312 | ||
AF125253.1 | - | 2 | 411 | |||
AK225422.1 | - | 2 | 327 | |||
AK290352.1 | - | 2 | 212 | |||
AK294750.1 | - | 2 | 190 | |||
AW163038.1 | - | 2 | 412 | |||
AY550966.1 | - | 2 | 212 | |||
AY698023.1 | - | 2 | 166 | |||
AY698024.1 | - | 2 | 166 | |||
BC094761.1 | - | 2 | 423 | 30528231 | ||
DQ088980.1 | - | 2 | 212 | |||
GU255993.1 | - | 2 | 209 | |||
HQ912715.1 | - | 2 | 412 | |||
K03193.1 | - | 2 | 325 | |||
U48722.1 | - | 2 | 410 | |||
U95089.1 | - | 3 | 669 | |||
X00588.1 | - | 2 | 352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
