siRNA Details
Gene Symbol: TNFRSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243.4 | NP_001234.3 | 11 | 1468 | ||
NM_001281430.2 | NP_001268359.2 | 10 | 1380 | |||
XM_011542441.2 | XP_011540743.1 | 11 | 1527 | |||
XM_011542443.2 | XP_011540745.1 | 10 | 1545 | |||
GenBank mRNA | BC063482.1 | - | 1 | 143 | ||
BC073839.1 | - | 1 | 110 | |||
BC136400.1 | - | 10 | 1287 | 9020387 | ||
BC141804.1 | - | 1 | 23 | |||
BC171762.1 | - | 10 | 1287 | |||
D86042.1 | - | 1 | 145 | |||
M83554.1 | - | 11 | 1468 | |||
S75768.1 | - | 11 | 1246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
