siRNA Details
Gene Symbol: CD40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001250.5 | NP_001241.1 | 4 | 380 | ||
NM_001302753.1 | NP_001289682.1 | 4 | 380 | |||
NM_001322421.1 | NP_001309350.1 | 4 | 380 | |||
NM_001322422.1 | NP_001309351.1 | 4 | 380 | |||
NM_152854.3 | NP_690593.1 | 4 | 380 | |||
NR_126502.1 | 4 | 380 | ||||
NR_136327.1 | 4 | 380 | ||||
XM_005260619.3 | XP_005260676.1 | 4 | 392 | |||
XM_011529109.2 | XP_011527411.1 | 4 | 392 | |||
XM_017028135.1 | XP_016883624.1 | 4 | 392 | |||
XM_017028136.1 | XP_016883625.1 | 4 | 391 | |||
XM_017028137.1 | XP_016883626.1 | 4 | 389 | |||
GenBank mRNA | AB209660.1 | - | 4 | 297 | ||
AJ300189.1 | - | 4 | 318 | |||
AK222896.1 | - | 4 | 320 | |||
AY225405.1 | - | 4 | 290 | |||
BC012419.1 | - | 4 | 347 | 3856903 | ||
BC064518.1 | - | 4 | 356 | 5227115 | ||
BM761221.1 | - | 4 | 367 | |||
BT019901.1 | - | 4 | 290 | |||
BX364285.2 | - | 4 | 334 | |||
HY063089.1 | - | 4 | 380 | |||
KU177966.1 | - | 4 | 290 | |||
KU177967.1 | - | 4 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
