siRNA Details
Gene Symbol: CXADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207063.1 | NP_001193992.1 | 4 | 659 | ||
NM_001207066.1 | NP_001193995.1 | 4 | 659 | |||
NM_001338.4 | NP_001329.1 | 4 | 659 | |||
XM_011529476.2 | XP_011527778.1 | 4 | 549 | |||
XM_011529477.2 | XP_011527779.1 | 4 | 549 | |||
XM_011529478.2 | XP_011527780.1 | 4 | 549 | |||
XM_011529479.1 | XP_011527781.1 | 4 | 549 | |||
GenBank mRNA | AF124598.1 | - | 4 | 435 | ||
AK313526.1 | - | 4 | 522 | |||
AY072912.1 | - | 4 | 435 | |||
BC003684.1 | - | 4 | 536 | 3463613 | ||
BC010536.1 | - | 4 | 510 | 3456544 | ||
BM547121.1 | - | 4 | 583 | |||
BT019876.1 | - | 4 | 435 | |||
GU474812.1 | - | 4 | 435 | |||
U90716.1 | - | 4 | 435 | |||
Y07593.1 | - | 4 | 494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
