siRNA Details
Gene Symbol: CHEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005735.1 | NP_001005735.1 | 4, 5 | 652 | ||
NM_001257387.1 | NP_001244316.1 | 3, 4 | 523 | |||
NM_007194.3 | NP_009125.1 | 3, 4 | 523 | |||
NM_145862.2 | NP_665861.1 | 3, 4 | 523 | |||
XM_011529839.2 | XP_011528141.1 | 4, 5 | 674 | |||
XM_011529840.2 | XP_011528142.1 | 4, 5 | 676 | |||
XM_011529844.2 | XP_011528146.1 | 4, 5 | 684 | |||
XM_011529845.2 | XP_011528147.1 | 2, 3 | 230 | |||
XM_017028560.1 | XP_016884049.1 | 4, 5 | 638 | |||
GenBank mRNA | AB040105.1 | - | 2, 3 | 451 | ||
AF086904.1 | - | 2, 3 | 451 | |||
AF096279.1 | - | 2, 3 | 451 | |||
AF174135.1 | - | 2, 3 | 451 | |||
AF217975.1 | - | 3, 4 | 1122 | |||
AJ131197.1 | - | 2, 3 | 451 | |||
AJ783839.1 | - | 2, 3 | 451 | |||
AK289360.1 | - | 3, 4 | 502 | |||
AK290754.1 | - | 3, 4 | 525 | |||
AY551297.1 | - | 3, 4 | 580 | |||
AY551298.1 | - | 2, 3 | 451 | |||
AY551299.1 | - | 2, 3 | 451 | |||
AY551300.1 | - | 2, 3 | 451 | |||
AY551302.1 | - | 2, 3 | 451 | |||
AY551305.1 | - | 2, 3 | 451 | |||
BC004207.2 | - | 3, 4 | 478 | 3532866 | ||
CR456418.1 | - | 4, 5 | 651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
