siRNA Details
Gene Symbol: GFRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001495.4 | NP_001486.4 | 2, 3 | 1037 | ||
XM_006716327.3 | XP_006716390.1 | 3, 4 | 483 | |||
XM_011544484.2 | XP_011542786.1 | 3, 4 | 515 | |||
GenBank mRNA | AF002700.1 | - | 2, 3 | 397 | ||
BC041688.1 | - | 2, 3 | 1037 | 5729102 | ||
U93703.1 | - | 2, 3 | 362 | |||
U97145.1 | - | 2, 3 | 615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
