siRNA Details
Gene Symbol: GFRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165038.1 | NP_001158510.1 | 7 | 1601 | ||
NM_001165039.1 | NP_001158511.1 | 6 | 1517 | |||
NM_001495.4 | NP_001486.4 | 8 | 1916 | |||
XM_006716327.3 | XP_006716390.1 | 9 | 1362 | |||
XM_011544484.2 | XP_011542786.1 | 9 | 1394 | |||
GenBank mRNA | AF002700.1 | - | 8 | 1276 | ||
AK299623.1 | - | 7 | 1007 | |||
AY326396.1 | - | 6 | 842 | |||
AY941828.1 | - | 7 | 926 | |||
BC041688.1 | - | 8 | 1916 | 5729102 | ||
U93703.1 | - | 8 | 1241 | |||
U97145.1 | - | 8 | 1494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
