siRNA Details
Gene Symbol: GFRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165038.1 | NP_001158510.1 | 8 | 1673 | ||
NM_001165039.1 | NP_001158511.1 | 7 | 1589 | |||
NM_001495.4 | NP_001486.4 | 9 | 1988 | |||
XM_006716327.3 | XP_006716390.1 | 10 | 1434 | |||
XM_011544484.2 | XP_011542786.1 | 10 | 1466 | |||
GenBank mRNA | AF002700.1 | - | 9 | 1348 | ||
AK299623.1 | - | 8 | 1079 | |||
AY326396.1 | - | 7 | 914 | |||
AY941828.1 | - | 8 | 998 | |||
BC041688.1 | - | 9 | 1988 | 5729102 | ||
U93703.1 | - | 9 | 1313 | |||
U97145.1 | - | 9 | 1566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
