siRNA Details
Gene Symbol: SCYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317784.1 | NP_001304713.1 | 18 | 2640 | ||
NM_017988.5 | NP_060458.3 | 18 | 2792 | |||
XM_017019618.1 | XP_016875107.1 | 19 | 2652 | |||
XM_017019619.1 | XP_016875108.1 | 19 | 2804 | |||
XM_017019620.1 | XP_016875109.1 | 20 | 2791 | |||
GenBank mRNA | AB037781.1 | - | 16 | 1958 | ||
AK001597.1 | - | 8 | 952 | |||
AK024274.1 | - | 14 | 1801 | |||
AK026160.1 | - | 1 | 57 | |||
AK027551.1 | - | 14 | 1858 | |||
AK292980.1 | - | 18 | 2566 | |||
BC012387.2 | - | 9 | 1085 | |||
BC063798.1 | - | 18 | 2405 | 6473586 |
