siRNA Details
Gene Symbol: RIOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159749.1 | NP_001153221.1 | 3 | 331 | ||
NM_018343.2 | NP_060813.2 | 3 | 331 | |||
GenBank mRNA | AK002021.1 | - | 3 | 270 | ||
AK225348.1 | - | 3 | 252 | |||
BC000953.2 | - | 3 | 259 | 3449997 | ||
DB240299.1 | - | 3 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
