siRNA Details
Gene Symbol: IL10RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001558.3 | NP_001549.2 | 2, 3 | 273 | ||
NR_026691.1 | 3, 4 | 403 | ||||
GenBank mRNA | AB209626.1 | - | 1, 2 | 1774 | ||
AK291645.1 | - | 2, 3 | 273 | |||
AK301484.1 | - | 3, 4 | 403 | |||
AK301492.1 | - | 2, 3 | 543 | |||
AK313049.1 | - | 2, 3 | 269 | |||
AK316146.1 | - | 2, 3 | 325 | |||
AK316342.1 | - | 3, 4 | 477 | |||
BC028082.1 | - | 2, 3 | 250 | 5216986 | ||
BM193545.1 | - | 3, 4 | 401 | |||
JN591377.1 | - | 2, 3 | 196 | |||
U00672.1 | - | 2, 3 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
