siRNA Details
Gene Symbol: IL10RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001558.3 | NP_001549.2 | 7 | 2000 | ||
NR_026691.1 | 8 | 2130 | ||||
GenBank mRNA | AB209626.1 | - | 6 | 3501 | ||
AK291645.1 | - | 7 | 2000 | |||
AK301484.1 | - | 8 | 2131 | |||
AK316146.1 | - | 7 | 2052 | |||
AK316342.1 | - | 8 | 2204 | |||
BC028082.1 | - | 7 | 1977 | 5216986 | ||
U00672.1 | - | 7 | 1980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
