siRNA Details
Gene Symbol: IL12RB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258214.1 | NP_001245143.1 | 7, 8 | 1530 | ||
NM_001258215.1 | NP_001245144.1 | 6, 7 | 1442 | |||
NM_001258216.1 | NP_001245145.1 | 6, 7 | 1442 | |||
NM_001319233.1 | NP_001306162.1 | 7, 8 | 1526 | |||
NM_001559.2 | NP_001550.1 | 6, 7 | 1442 | |||
NR_047583.1 | 6, 7 | 1442 | ||||
NR_047584.1 | 6, 7 | 1442 | ||||
XM_005270825.2 | XP_005270882.1 | 7, 8 | 1543 | |||
XM_005270827.2 | XP_005270884.1 | 7, 8 | 1539 | |||
XM_005270828.3 | XP_005270885.1 | 6, 7 | 1451 | |||
XM_006710617.2 | XP_006710680.1 | 7, 8 | 1543 | |||
XM_011541383.2 | XP_011539685.1 | 7, 8 | 995 | |||
XM_011541384.2 | XP_011539686.1 | 7, 8 | 1543 | |||
XM_017001202.1 | XP_016856691.1 | 7, 8 | 1543 | |||
XM_017001203.1 | XP_016856692.1 | 6, 7 | 1455 | |||
XM_017001204.1 | XP_016856693.1 | 7, 8 | 1543 | |||
GenBank mRNA | AK294494.1 | - | 7, 8 | 1021 | ||
AK316525.1 | - | 7, 8 | 1001 | |||
BC104772.1 | - | 5, 6 | 852 | 8143775 | ||
BC104774.1 | - | 5, 6 | 852 | 8143777 | ||
BC143248.1 | - | 5, 6 | 852 | |||
BC143249.1 | - | 5, 6 | 852 | 9051751 | ||
BC143250.1 | - | 5, 6 | 852 | |||
U64198.1 | - | 6, 7 | 1442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
