siRNA Details
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001560.2 | NP_001551.1 | 2 | 219 | ||
XM_011531336.1 | XP_011529638.1 | 2 | 497 | |||
XM_017029507.1 | XP_016884996.1 | 2 | 375 | |||
GenBank mRNA | AB209849.1 | - | 2 | 174 | ||
AK313467.1 | - | 2 | 208 | |||
BC009960.2 | - | 2 | 206 | 4300487 | ||
BC015768.1 | - | 2 | 223 | 4868206 | ||
U81379.3 | - | 2 | 216 | |||
U81380.2 | - | 2 | 216 | |||
Y09328.1 | - | 2 | 209 | |||
Y10659.1 | - | 2 | 219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
