siRNA Details
Gene Symbol: TNFRSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001561.5 | NP_001552.2 | 4 | 448 | ||
XM_006710618.3 | XP_006710681.1 | 3 | 1095 | |||
XM_011541386.2 | XP_011539688.1 | 3 | 463 | |||
GenBank mRNA | BC006196.2 | - | 3 | 308 | 2924109 | |
BP381234.1 | - | 4 | 448 | |||
L12964.2 | - | 3 | 326 | |||
U03397.1 | - | 3 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
