siRNA Details
Gene Symbol: HIPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113239.2 | NP_001106710.1 | 5, 6 | 1713 | ||
NM_022740.4 | NP_073577.3 | 5, 6 | 1713 | |||
XM_006715935.2 | XP_006715998.1 | 6, 7 | 1702 | |||
XM_011516077.2 | XP_011514379.1 | 5, 6 | 2291 | |||
XM_011516078.2 | XP_011514380.1 | 5, 6 | 2292 | |||
XM_011516079.2 | XP_011514381.1 | 5, 6 | 2291 | |||
XM_011516080.2 | XP_011514382.1 | 5, 6 | 2292 | |||
XM_011516081.2 | XP_011514383.1 | 5, 6 | 1688 | |||
XM_017012069.1 | XP_016867558.1 | 5, 6 | 1794 | |||
GenBank mRNA | AF207702.1 | - | 4, 5 | 1421 | ||
AF208291.1 | - | 5, 6 | 1550 | |||
AF326592.1 | - | 5, 6 | 1597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
