siRNA Details
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK223393.1 | - | 2 | 343 | ||
AK301154.1 | - | 2 | 366 | |||
AK303367.1 | - | 2 | 339 | |||
AK313614.1 | - | 2 | 372 | |||
BC033583.1 | - | 2 | 327 | 5227138 | ||
CD691699.1 | - | 2 | 397 | |||
CR996443.1 | - | 2 | 349 | |||
M14362.1 | - | 2 | 323 | |||
M16445.1 | - | 2 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
