siRNA Details
Gene Symbol: COASY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042529.2 | NP_001035994.1 | 2 | 342 | ||
NM_001042532.3 | NP_001035997.2 | 3 | 474 | |||
NM_025233.6 | NP_079509.5 | 1 | 734 | |||
XM_006722116.3 | XP_006722179.1 | 2 | 368 | |||
XM_011525300.1 | XP_011523602.1 | 2 | 538 | |||
XM_017025167.1 | XP_016880656.1 | 1 | 678 | |||
GenBank mRNA | AF453478.1 | - | 1 | 185 | ||
AK075415.1 | - | 1 | 641 | |||
AK225485.1 | - | 2 | 271 | |||
AK297153.1 | - | 3 | 397 | |||
AK303766.1 | - | 2 | 265 | |||
AK314076.1 | - | 3 | 398 | |||
AL360168.1 | - | 2 | 516 | |||
AL583142.3 | - | 2 | 260 | |||
AL833588.1 | - | 2 | 266 | |||
AY094602.1 | - | 2 | 295 | |||
BC067254.1 | - | 1 | 638 | 6502943 | ||
BM473635.1 | - | 2 | 515 | |||
BM781531.1 | - | 2 | 521 | |||
DA786254.1 | - | 2 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
