siRNA Details
Gene Symbol: FCER1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002001.3 | NP_001992.1 | 6 | 775 | ||
GenBank mRNA | BC005912.1 | - | 5 | 698 | 4294467 | |
BC015195.1 | - | 5 | 702 | 4251469 | ||
J03605.1 | - | 5 | 703 | |||
X06948.1 | - | 6 | 775 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
