siRNA Details
Gene Symbol: FNTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002027.2 | NP_002018.1 | 3 | 402 | ||
NR_033698.1 | 2 | 316 | ||||
GenBank mRNA | AK123002.1 | - | 3 | 789 | ||
AK292121.1 | - | 3 | 378 | |||
BC017029.2 | - | 3 | 345 | |||
BC084566.1 | - | 3 | 362 | 6423445 | ||
BI913631.1 | - | 3 | 549 | |||
BM686906.1 | - | 2 | 295 | |||
BT009854.1 | - | 3 | 354 | |||
L00634.1 | - | 3 | 370 | |||
L10413.1 | - | 3 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
