siRNA Details
Gene Symbol: EIF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267810.1 | NP_001254739.1 | 4, 5 | 402 | ||
NM_002212.3 | NP_002203.1 | 4, 5 | 472 | |||
NM_181468.2 | NP_852133.1 | 3, 4 | 606 | |||
NR_052023.1 | 4, 5 | 465 | ||||
GenBank mRNA | AB062289.1 | - | 4, 5 | 405 | ||
AF022229.1 | - | 3, 4 | 461 | |||
AF047433.1 | - | 4, 5 | 446 | |||
AK314969.1 | - | 4, 5 | 448 | |||
AL537456.3 | - | 3, 4 | 599 | |||
BC001119.2 | - | 4, 5 | 445 | 2989973 | ||
BC011845.2 | - | 4, 5 | 423 | 4120463 | ||
BC019305.1 | - | 4, 5 | 405 | 2823211 | ||
BQ222221.1 | - | 3, 4 | 392 | |||
CR456764.1 | - | 3, 4 | 376 | |||
CT002623.1 | - | 4, 5 | 417 | |||
Y11435.1 | - | 4, 5 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
