siRNA Details
Gene Symbol: IL1RAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167928.1 | NP_001161400.1 | 7, 8 | 1125 | ||
NM_001167929.1 | NP_001161401.1 | 5, 6 | 874 | |||
NM_001167930.1 | NP_001161402.1 | 5, 6 | 874 | |||
NM_001167931.1 | NP_001161403.1 | 6, 7 | 961 | |||
NM_002182.3 | NP_002173.1 | 6, 7 | 961 | |||
NM_134470.3 | NP_608273.1 | 6, 7 | 961 | |||
XM_017006347.1 | XP_016861836.1 | 5, 6 | 874 | |||
XM_017006348.1 | XP_016861837.1 | 2, 3 | 275 | |||
GenBank mRNA | AB006537.1 | - | 6, 7 | 910 | ||
AF029213.1 | - | 4, 5 | 713 | |||
AF167343.1 | - | 4, 5 | 704 | |||
AF538730.1 | - | 6, 7 | 910 | |||
AF538731.1 | - | 5, 6 | 823 | |||
AF538732.1 | - | 5, 6 | 823 | |||
AF538733.1 | - | 5, 6 | 823 | |||
AF538734.1 | - | 6, 7 | 910 | |||
AK291669.1 | - | 7, 8 | 1120 | |||
BC053621.1 | - | 6, 7 | 914 | 6160894 | ||
BM760155.1 | - | 2, 3 | 238 | |||
EF591790.1 | - | 6, 7 | 910 | |||
FJ998418.1 | - | 4, 5 | 704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
