siRNA Details
Gene Symbol: IL6ST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190981.1 | NP_001177910.1 | 6 | 939 | ||
NM_002184.3 | NP_002175.2 | 6 | 939 | |||
NM_175767.2 | NP_786943.1 | 6 | 939 | |||
NR_120480.1 | 6 | 939 | ||||
XM_017009441.1 | XP_016864930.1 | 5 | 795 | |||
XM_017009442.1 | XP_016864931.1 | 5 | 818 | |||
GenBank mRNA | AB015706.1 | - | 5 | 718 | ||
AB102802.1 | - | 4 | 626 | |||
BC117402.1 | - | 5 | 738 | 40125953 | ||
BC117404.1 | - | 5 | 738 | 40125955 | ||
HQ628629.1 | - | 4 | 626 | |||
HQ628630.1 | - | 4 | 626 | |||
M57230.1 | - | 6 | 881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
